| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500018140.mm.1 |
NM_001038658.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500018140.mm.1 |
NM_028224.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500018140.mm.1 |
TC1500002214.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500018139.mm.1 |
NM_001038658.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500018139.mm.1 |
NM_028224.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500018139.mm.1 |
TC1500002214.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500018142.mm.1 |
NM_001038658.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500018142.mm.1 |
NM_028224.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500018142.mm.1 |
TC1500002214.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500018141.mm.1 |
NM_001038658.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500018141.mm.1 |
NM_028224.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500018141.mm.1 |
TC1500002214.mm.1 |
MTA TC |