Assay Details
| Gene Symbol: Ccnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052836.1 | - | 87 | |||
| AK076134.1 | - | 87 | ||||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnt1| Gene Symbol: Ccnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052836.1 | - | 87 | |||
| AK076134.1 | - | 87 | ||||
Set Membership: |
|