Assay Details
| Gene Symbol: Adam4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009620.1 | NP_033750.1 | 1 | 876 | 63 | |
| GenBank mRNA | AY158688.1 | - | 1 | 876 | 63 | |
| BC150159.1 | - | 1 | 880 | 40141514 | 63 | |
| BC150481.1 | - | 1 | 880 | 40141515 | 63 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101707 | BC150481.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101707 | AY158688.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101707 | BC150159.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101707 | NM_009620.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101707 | AK076647.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232860 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 54440 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101707 | BC150481.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101707 | AY158688.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101707 | BC150159.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101707 | NM_009620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101707 | AK076647.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02560649 | - | - |
| Whole Mouse Genome G4122A | A_52_P462447 | BC150481.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462447 | AY158688.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462447 | BC150159.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462447 | NM_009620.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462447 | AK076647.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101707 | BC150481.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101707 | AY158688.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101707 | BC150159.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101707 | NM_009620.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101707 | AK076647.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02560649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28581081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10249008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28581105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10339509 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10579994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401321 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427502_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10579993 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10401320 | - | - |
| Mouse Genome 430 2.0 Array | 1427502_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427502_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa16250_at | - | - |
| Mouse Expression Set 430 | 1427502_at | - | - |
| Mouse Expression Set 430 | 1427502_PM_at | - | - |
| Murine Genome U74v2 Set | 92733_PM_at | - | - |
| Murine Genome U74v2 Set | 92733_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427502_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427502_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427502_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427502_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 409207 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6200372 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 630577 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6200372 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000630577 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200372 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249290 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235132 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249290 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235132 | - | - |
| mouse-6 v1.1 (Illumina) | scl0011498.2_120-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0011498.2_254-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200372 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 630577 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0011498.2_254-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0011498.2_120-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200372 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630577 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1235132 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1249290 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2971964 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2971964 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235132 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249290 | - | - |

Adam4
Genomic Map