Assay Details
| Gene Symbol: Mapk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011950.2 | NP_036080.2 | 11 - 12 | 1110 | 94 | |
| GenBank mRNA | AK146993.1 | - | 11 - 12 | 1098 | 94 | |
| BC001992.1 | - | 11 - 12 | 1079 | 3492903 | 94 | |
| U81823.1 | - | 11 - 12 | 1067 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
response to stress
response to osmotic stress
cell cycle
phosphorylation
peptidyl-serine phosphorylation
positive regulation of interleukin-6 production
intracellular signal transduction
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
MAPK cascade
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
response to stress
response to osmotic stress
cell cycle
phosphorylation
peptidyl-serine phosphorylation
positive regulation of interleukin-6 production
intracellular signal transduction
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31124214 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31124213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31124210 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31124209 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31124215 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239203 | AK146993.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239203 | U81823.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239203 | NM_011950.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239203 | AK146993.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239203 | U81823.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239203 | NM_011950.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646931 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896271 | AF029748.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 24814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675472 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166833 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239203 | AK146993.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239203 | U81823.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239203 | NM_011950.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239203 | AK146993.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239203 | U81823.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239203 | NM_011950.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239203 | AK146993.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239203 | U81823.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239203 | NM_011950.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239203 | AK146993.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239203 | U81823.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239203 | NM_011950.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31124220 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166833 | - | - |
| Whole Mouse Genome G4122A | A_51_P239203 | AK146993.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239203 | U81823.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239203 | NM_011950.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239203 | AK146993.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239203 | U81823.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239203 | NM_011950.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130382 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166833 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13247422 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13522219 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13479290 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13522216 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31124213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31124209 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31124215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31124210 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31124214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31124191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13086433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13522218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13479293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13065138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13105904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13437878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13522216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13535882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13093798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13562704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13535879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13537634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13425516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13309206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13105903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13266363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13222268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31124203 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10443415 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443409 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443410 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443412 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443416 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443414 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443413 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443411 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443417 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448871_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10443408 | - | - |
| Mouse Genome 430 2.0 Array | 1448871_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448871_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26415_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26415_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700004240.mm.1 | NM_011950.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004240.mm.1 | NM_011950.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004240.mm.1 | TC1700000487.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700004242.mm.1 | NM_011950.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004242.mm.1 | NM_011950.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004242.mm.1 | TC1700000487.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700004241.mm.1 | NM_011950.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004241.mm.1 | NM_011950.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004241.mm.1 | TC1700000487.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448871_PM_at | - | - |
| Mouse Expression Set 430 | 1448871_at | - | - |
| Murine Genome U74v2 Set | 160763_PM_at | - | - |
| Murine Genome U74v2 Set | 160763_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448871_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448871_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448871_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448871_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 499918 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6760215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6760215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006760215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1212692 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1212692 | - | - |
| mouse-6 v1.1 (Illumina) | scl50917.9.3_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6760215 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50917.9.3_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760215 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2955452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1212692 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1212692 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2955452 | - | - |

Mapk13
Genomic Map