Assay Details
| Gene Symbol: Mapk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011950.2 | NP_036080.2 | 2 - 3 | 344 | 97 | |
| GenBank mRNA | AK146993.1 | - | 2 - 3 | 332 | 97 | |
| BC001992.1 | - | 2 - 3 | 313 | 3492903 | 97 | |
| CJ182359.1 | - | 2 - 3 | 346 | 97 | ||
| U81823.1 | - | 2 - 3 | 301 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk13
Genomic Map