Assay Details
| Gene Symbol: Srpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016795.3 | NP_058075.2 | 1 - 2 | 52 | 76 | |
| GenBank mRNA | AK017187.1 | - | 1 - 2 | 49 | 76 | |
| AK141271.1 | - | 1 - 2 | 119 | 76 | ||
| AK151152.1 | - | 1 - 2 | 115 | 76 | ||
| BC050761.1 | - | 1 - 2 | 52 | 6771151 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK141271.1,AK151152.1,BC050761.1,NM_016795.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Srpk1
Genomic Map