Assay Details
| Gene Symbol: Srpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016795.3 | NP_058075.2 | 3 - 4 | 225 | 97 | |
| GenBank mRNA | AB012290.1 | - | 2 - 3 | 183 | 97 | |
| AJ224115.1 | - | 3 - 4 | 219 | 97 | ||
| AK017187.1 | - | 3 - 4 | 222 | 97 | ||
| AK141271.1 | - | 3 - 4 | 292 | 97 | ||
| AK151152.1 | - | 3 - 4 | 288 | 97 | ||
| AK161346.1 | - | 3 - 4 | 203 | 97 | ||
| AK161388.1 | - | 3 - 4 | 204 | 97 | ||
| BC005707.1 | - | 3 - 4 | 194 | 3157835 | 97 | |
| BC050761.1 | - | 3 - 4 | 225 | 6771151 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srpk1
Genomic Map