Assay Details
| Gene Symbol: Mapk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168513.1 | NP_001161985.1 | 8 - 9 | 675 | 77 | |
| NM_001168514.1 | NP_001161986.1 | 9 - 10 | 1119 | 77 | ||
| NM_011951.3 | NP_036081.1 | 8 - 9 | 1074 | 77 | ||
| GenBank mRNA | AF128892.1 | - | 8 - 9 | 972 | 77 | |
| AK133684.1 | - | 8 - 9 | 987 | 77 | ||
| AK151348.1 | - | 8 - 9 | 676 | 77 | ||
| AK153025.1 | - | 8 - 9 | 676 | 77 | ||
| BC012235.1 | - | 8 - 9 | 1074 | 4195842 | 77 | |
| BC018227.1 | - | 9 - 10 | 1016 | 77 | ||
| U10871.1 | - | 8 - 9 | 972 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk14
Genomic Map