Assay Details
| Gene Symbol: Msln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018857.1 | NP_061345.1 | 11 - 12 | 1348 | 65 | |
| GenBank mRNA | AK144391.1 | - | 12 - 13 | 1280 | 65 | |
| AK146799.1 | - | 11 - 12 | 1398 | 65 | ||
| BC023753.1 | - | 11 - 12 | 1352 | 5354484 | 65 | |
| D86370.1 | - | 11 - 12 | 1348 | 65 | ||
More Information
Set Membership: |
|

Msln
Genomic Map