Assay Details
| Gene Symbol: Narfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026238.4 | NP_080514.3 | 2 - 3 | 210 | 60 | |
| GenBank mRNA | AK014038.1 | - | 2 - 3 | 194 | 60 | |
| AK018548.1 | - | 2 - 3 | 181 | 60 | ||
| AK043694.1 | - | 2 - 3 | 187 | 60 | ||
| AK145408.1 | - | 2 - 3 | 213 | 60 | ||
| BC020468.1 | - | 2 - 3 | 170 | 60 | ||
| BC025108.1 | - | 2 - 3 | 190 | 60 | ||
| BC052830.1 | - | 2 - 3 | 187 | 30040958 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Narfl
Genomic Map