Assay Details
| Gene Symbol: Narfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026238.4 | NP_080514.3 | 7 - 8 | 868 | 74 | |
| GenBank mRNA | AK003464.1 | - | 2 - 3 | 201 | 74 | |
| AK014038.1 | - | 7 - 8 | 852 | 74 | ||
| AK018548.1 | - | 7 - 8 | 839 | 74 | ||
| AK043694.1 | - | 7 - 8 | 845 | 74 | ||
| AK145408.1 | - | 7 - 8 | 871 | 74 | ||
| BC020468.1 | - | 7 - 8 | 828 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Narfl
Genomic Map