Assay Details
| Gene Symbol: Haghl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271433.1 | NP_001258362.1 | 1 - 2 | 159 | 94 | |
| NM_001271434.1 | NP_001258363.1 | 1 - 2 | 159 | 94 | ||
| NM_001271435.1 | NP_001258364.1 | 1 - 2 | 159 | 94 | ||
| GenBank mRNA | AK012748.1 | - | 1 - 2 | 156 | 94 | |
| AK034798.1 | - | 1 - 2 | 134 | 94 | ||
| AK047517.1 | - | 1 - 2 | 114 | 94 | ||
| BC005586.1 | - | 1 - 2 | 105 | 94 | ||
| BC030466.1 | - | 1 - 2 | 144 | 5253213 | 94 | |
| BC083322.1 | - | 1 - 2 | 98 | 6820885 | 94 | |
| BY265614.1 | - | 1 - 2 | 159 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK047517.1, BC030466.1, BC083322.1, NM_001271433.1, NM_001271434.1, NM_001271435.1 transcripts.
Set Membership: |
|

Haghl
Genomic Map