Assay Details
| Gene Symbol: Haghl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271433.1 | NP_001258362.1 | 3 - 4 | 421 | 75 | |
| NM_001271434.1 | NP_001258363.1 | 3 - 4 | 421 | 75 | ||
| NM_001271435.1 | NP_001258364.1 | 3 - 4 | 421 | 75 | ||
| NM_001271436.1 | NP_001258365.1 | 2 - 3 | 235 | 75 | ||
| NM_001271437.1 | NP_001258366.1 | 2 - 3 | 230 | 75 | ||
| NM_026897.3 | NP_081173.1 | 2 - 3 | 406 | 75 | ||
| GenBank mRNA | AK005274.1 | - | 2 - 3 | 402 | 75 | |
| AK012748.1 | - | 3 - 4 | 418 | 75 | ||
| AK021220.1 | - | 2 - 3 | 232 | 75 | ||
| AK032999.1 | - | 2 - 3 | 212 | 75 | ||
| AK034798.1 | - | 3 - 4 | 396 | 75 | ||
| AK047517.1 | - | 3 - 4 | 376 | 75 | ||
| AK079632.1 | - | 2 - 3 | 404 | 75 | ||
| BC005586.1 | - | 3 - 4 | 367 | 75 | ||
| BC030466.1 | - | 3 - 4 | 406 | 5253213 | 75 | |
| BC083322.1 | - | 3 - 4 | 360 | 6820885 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271436.1, NM_001271437.1 transcripts.
Set Membership: |
|

Haghl
Genomic Map