Assay Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042673.2 | NP_001036138.1 | 10 - 11 | 1331 | 88 | |
| NM_001282942.1 | NP_001269871.1 | 9 - 10 | 1134 | 88 | ||
| NM_007985.3 | NP_032011.2 | 10 - 11 | 1331 | 88 | ||
| GenBank mRNA | AK035488.1 | - | 10 - 11 | 1085 | 88 | |
| AK042226.1 | - | 10 - 11 | 1301 | 88 | ||
| AK080697.1 | - | 1312 | 88 | |||
| AK082717.1 | - | 9 - 10 | 1112 | 88 | ||
| AK147025.1 | - | 10 - 11 | 1311 | 88 | ||
| BC038609.1 | - | 9 - 10 | 1116 | 5375091 | 88 | |
| L08266.1 | - | 10 - 11 | 1312 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fancc
Genomic Map