Assay Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042673.2 | NP_001036138.1 | 1 - 2 | 301 | 96 | |
| NM_007985.3 | NP_032011.2 | 1 - 2 | 301 | 96 | ||
| GenBank mRNA | AK080697.1 | - | 281 | 96 | ||
| AK147025.1 | - | 1 - 2 | 281 | 96 | ||
| AK163161.1 | - | 1 - 2 | 302 | 96 | ||
| L08266.1 | - | 1 - 2 | 282 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the L08266.1, NM_001042673.2, NM_007985.3 transcripts.
Set Membership: |
|

Fancc
Genomic Map