Assay Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042673.2 | NP_001036138.1 | 3 - 4 | 576 | 132 | |
| NM_001282942.1 | NP_001269871.1 | 2 - 3 | 379 | 132 | ||
| NM_007985.3 | NP_032011.2 | 3 - 4 | 576 | 132 | ||
| GenBank mRNA | AK035488.1 | - | 3 - 4 | 330 | 132 | |
| AK080697.1 | - | 556 | 133 | |||
| AK082717.1 | - | 2 - 3 | 357 | 132 | ||
| AK147025.1 | - | 3 - 4 | 556 | 132 | ||
| AK163161.1 | - | 3 - 4 | 577 | 132 | ||
| BC038609.1 | - | 2 - 3 | 361 | 5375091 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK082717.1, BC038609.1, NM_001282942.1 transcripts.
Set Membership: |
|

Fancc
Genomic Map