Assay Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042673.2 | NP_001036138.1 | 4 - 5 | 672 | 96 | |
| NM_001282942.1 | NP_001269871.1 | 3 - 4 | 475 | 96 | ||
| NM_007985.3 | NP_032011.2 | 4 - 5 | 672 | 96 | ||
| GenBank mRNA | AK035488.1 | - | 4 - 5 | 426 | 96 | |
| AK042226.1 | - | 4 - 5 | 642 | 96 | ||
| AK080697.1 | - | 653 | 96 | |||
| AK082717.1 | - | 3 - 4 | 453 | 96 | ||
| AK147025.1 | - | 4 - 5 | 652 | 96 | ||
| AK163161.1 | - | 4 - 5 | 673 | 96 | ||
| BC038609.1 | - | 3 - 4 | 457 | 5375091 | 96 | |
| L08266.1 | - | 4 - 5 | 653 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC038609.1 transcripts.
Set Membership: |
|

Fancc
Genomic Map