Assay Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042673.2 | NP_001036138.1 | 6 - 7 | 853 | 83 | |
| NM_001282942.1 | NP_001269871.1 | 5 - 6 | 656 | 83 | ||
| NM_007985.3 | NP_032011.2 | 6 - 7 | 853 | 83 | ||
| GenBank mRNA | AK035488.1 | - | 6 - 7 | 607 | 83 | |
| AK042226.1 | - | 6 - 7 | 823 | 83 | ||
| AK080697.1 | - | 834 | 83 | |||
| AK082717.1 | - | 5 - 6 | 634 | 83 | ||
| AK147025.1 | - | 6 - 7 | 833 | 83 | ||
| AK163161.1 | - | 6 - 7 | 854 | 83 | ||
| BC038609.1 | - | 5 - 6 | 638 | 5375091 | 83 | |
| L08266.1 | - | 6 - 7 | 834 | 83 | ||
More Information
Set Membership: |
|

Fancc
Genomic Map