Assay Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042673.2 | NP_001036138.1 | 8 - 9 | 1173 | 81 | |
| NM_001282942.1 | NP_001269871.1 | 7 - 8 | 976 | 81 | ||
| NM_007985.3 | NP_032011.2 | 8 - 9 | 1173 | 81 | ||
| GenBank mRNA | AK035488.1 | - | 8 - 9 | 927 | 81 | |
| AK042226.1 | - | 8 - 9 | 1143 | 81 | ||
| AK080697.1 | - | 1154 | 81 | |||
| AK082717.1 | - | 7 - 8 | 954 | 81 | ||
| AK147025.1 | - | 8 - 9 | 1153 | 81 | ||
| AK163161.1 | - | 8 - 9 | 1174 | 81 | ||
| BC038609.1 | - | 7 - 8 | 958 | 5375091 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fancc
Genomic Map