Assay Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042673.2 | NP_001036138.1 | 9 - 10 | 1228 | 65 | |
| NM_001282942.1 | NP_001269871.1 | 8 - 9 | 1031 | 65 | ||
| NM_007985.3 | NP_032011.2 | 9 - 10 | 1228 | 65 | ||
| GenBank mRNA | AK035488.1 | - | 9 - 10 | 982 | 65 | |
| AK042226.1 | - | 9 - 10 | 1198 | 65 | ||
| AK080697.1 | - | 1209 | 65 | |||
| AK082717.1 | - | 8 - 9 | 1009 | 65 | ||
| AK147025.1 | - | 9 - 10 | 1208 | 65 | ||
| BC038609.1 | - | 8 - 9 | 1013 | 5375091 | 65 | |
More Information
Set Membership: |
|

Fancc
Genomic Map