Assay Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016843.3 | NP_058539.2 | 6 - 7 | 815 | 85 | |
| GenBank mRNA | AK002504.1 | - | 6 - 7 | 924 | 85 | |
| AK003530.1 | - | 6 - 7 | 742 | 85 | ||
| AK047170.1 | - | 6 - 7 | 865 | 85 | ||
| AK049706.1 | - | 6 - 7 | 859 | 85 | ||
| AK077738.1 | - | 6 - 7 | 817 | 86 | ||
| AK078459.1 | - | 6 - 7 | 895 | 85 | ||
| AK153271.1 | - | 6 - 7 | 817 | 85 | ||
| AK161950.1 | - | 832 | 85 | |||
| AK165941.1 | - | 6 - 7 | 840 | 85 | ||
| AK166899.1 | - | 6 - 7 | 900 | 85 | ||
| AK170507.1 | - | 6 - 7 | 832 | 85 | ||
| AK170751.1 | - | 6 - 7 | 841 | 85 | ||
| AK195052.1 | - | 1 - 2 | 49 | 85 | ||
| BC016410.1 | - | 6 - 7 | 825 | 3968016 | 85 | |
| BC046802.1 | - | 6 - 7 | 951 | 6404607 | 85 | |
| X61506.1 | - | 6 - 7 | 929 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atxn10
Genomic Map