| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300017706.mm.1 |
NM_172807.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017706.mm.1 |
NM_172807.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017706.mm.1 |
TC1300002597.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017707.mm.1 |
NM_172807.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017707.mm.1 |
NM_172807.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017707.mm.1 |
TC1300002597.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017701.mm.1 |
NM_172807.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017701.mm.1 |
NM_172807.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017701.mm.1 |
TC1300002597.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017704.mm.1 |
NM_172807.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017704.mm.1 |
NM_172807.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017704.mm.1 |
TC1300002597.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017702.mm.1 |
NM_172807.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017702.mm.1 |
NM_172807.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017702.mm.1 |
TC1300002597.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017703.mm.1 |
NM_172807.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017703.mm.1 |
NM_172807.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017703.mm.1 |
TC1300002597.mm.1 |
MTA TC |