Assay Details
| Gene Symbol: Ppwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172807.4 | NP_766395.2 | 2 - 3 | 335 | 82 | |
| GenBank mRNA | AK043316.1 | - | 2 - 3 | 335 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppwd1| Gene Symbol: Ppwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172807.4 | NP_766395.2 | 2 - 3 | 335 | 82 | |
| GenBank mRNA | AK043316.1 | - | 2 - 3 | 335 | 82 | |
Set Membership: |
|