Assay Details
| Gene Symbol: Chrng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009604.3 | NP_033734.3 | 9 - 10 | 1051 | 108 | |
| GenBank mRNA | AK047974.1 | - | 9 - 10 | 1052 | 108 | |
| M30514.1 | - | 9 - 10 | 1092 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chrng
Genomic Map