Assay Details
| Gene Symbol: Chrng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009604.3 | NP_033734.3 | 7 - 8 | 61 | ||
| GenBank mRNA | AK047974.1 | - | 7 - 8 | 61 | ||
| M30514.1 | - | 7 - 8 | 859 | 61 | ||
| X03818.1 | - | 7 - 8 | 815 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chrng
Genomic Map