Assay Details
| Gene Symbol: Igsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207205.2 | NP_997088.2 | 5 - 6 | 1946 | 57 | |
| GenBank mRNA | AK045721.1 | - | 2 - 3 | 676 | 57 | |
| AK084011.1 | - | 5 - 6 | 1984 | 57 | ||
| AK129151.1 | - | 5 - 6 | 1946 | 57 | ||
| BC052892.1 | - | 4 - 5 | 1370 | 30055632 | 57 | |
More Information
Set Membership: |
|

Igsf3
Genomic Map