Assay Details
| Gene Symbol: Cdc14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122989.1 | NP_001116461.1 | 2 - 3 | 231 | 93 | |
| NM_172587.3 | NP_766175.3 | 2 - 3 | 495 | 93 | ||
| GenBank mRNA | AK041155.1 | - | 2 - 3 | 217 | 93 | |
| AK044332.1 | - | 2 - 3 | 797 | 93 | ||
| AK161345.1 | - | 2 - 3 | 496 | 93 | ||
| BC049794.1 | - | 2 - 3 | 437 | 6514098 | 93 | |
| BC057357.1 | - | 2 - 3 | 386 | 6835123 | 93 | |
| CN720173.1 | - | 2 - 3 | 149 | 93 | ||
More Information
Set Membership: |
|

Cdc14b
Genomic Map