Assay Details
| Gene Symbol: Cdc14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122989.1 | NP_001116461.1 | 7 - 8 | 602 | 114 | |
| NM_172587.3 | NP_766175.3 | 7 - 8 | 866 | 114 | ||
| GenBank mRNA | AK041155.1 | - | 7 - 8 | 588 | 114 | |
| AK044332.1 | - | 7 - 8 | 1168 | 114 | ||
| BC049794.1 | - | 7 - 8 | 808 | 6514098 | 114 | |
| BC057357.1 | - | 7 - 8 | 757 | 6835123 | 114 | |
| CN720173.1 | - | 7 - 8 | 520 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc14b
Genomic Map