Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100034565.mm.1 |
NM_010404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034565.mm.1 |
NM_010404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034565.mm.1 |
NM_177981.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034565.mm.1 |
NM_177981.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034565.mm.1 |
TC1100003766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034559.mm.1 |
NM_010404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034559.mm.1 |
NM_010404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034559.mm.1 |
NM_177981.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034559.mm.1 |
NM_177981.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034559.mm.1 |
TC1100003766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034561.mm.1 |
NM_010404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034561.mm.1 |
NM_010404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034561.mm.1 |
NM_177981.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034561.mm.1 |
NM_177981.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034561.mm.1 |
TC1100003766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034564.mm.1 |
NM_010404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034564.mm.1 |
NM_010404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034564.mm.1 |
NM_177981.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034564.mm.1 |
NM_177981.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034564.mm.1 |
TC1100003766.mm.1 |
MTA TC |