Assay Details
| Gene Symbol: Tnfrsf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011610.3 | NP_035740.2 | 6 - 7 | 860 | 62 | |
| GenBank mRNA | AK004752.1 | - | 6 - 7 | 861 | 62 | |
| AK004844.2 | - | 6 - 7 | 860 | 62 | ||
| AK140332.1 | - | 6 - 7 | 861 | 62 | ||
| AK153301.1 | - | 6 - 7 | 861 | 62 | ||
| AK155385.1 | - | 6 - 7 | 861 | 62 | ||
| BC141405.1 | - | 6 - 7 | 847 | 9056018 | 62 | |
| M59378.1 | - | 6 - 7 | 833 | 62 | ||
| M60469.1 | - | 6 - 7 | 831 | 62 | ||
| X76401.1 | - | 6 - 7 | 746 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnfrsf1b
Genomic Map