Assay Details
| Gene Symbol: Synrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194341.2 | NP_919322.1 | 7 - 8 | 700 | 85 | |
| GenBank mRNA | BC056370.1 | - | 7 - 8 | 660 | 6831893 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

Synrg
Genomic Map