Assay Details
| Gene Symbol: Rbmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166623.1 | NP_001160095.1 | 7 - 8 | 990 | 70 | |
| NM_011252.4 | NP_035382.1 | 7 - 8 | 987 | 70 | ||
| NR_029425.1 | 7 - 8 | 987 | 70 | |||
| GenBank mRNA | AJ237846.1 | - | 7 - 8 | 917 | 70 | |
| AJ237847.1 | - | 7 - 8 | 932 | 70 | ||
| AK083899.1 | - | 7 - 8 | 988 | 70 | ||
| AK087940.1 | - | 1 - 2 | 91 | 70 | ||
| AK135164.1 | - | 7 - 8 | 992 | 70 | ||
| AK165006.1 | - | 7 - 8 | 933 | 70 | ||
| AK178027.1 | - | 3 - 4 | 329 | 70 | ||
| AK211002.1 | - | 2 - 3 | 202 | 70 | ||
| BC003710.1 | - | 7 - 8 | 985 | 3599678 | 70 | |
More Information
Set Membership: |
|

Rbmx
Genomic Map