Assay Details
| Gene Symbol: Rbmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029425.1 | 9 - 10 | 1162 | 90 | ||
| GenBank mRNA | AJ237846.1 | - | 9 - 10 | 1092 | 90 | |
| AK087940.1 | - | 3 - 4 | 266 | 90 | ||
| AK165006.1 | - | 9 - 10 | 1108 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbmx
Genomic Map