Assay Details
| Gene Symbol: F9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK149372.1 | - | 5 - 6 | 523 | 105 | |
| BC125617.1 | - | 5 - 6 | 521 | 105 | ||
| BC132393.1 | - | 5 - 6 | 521 | 105 | ||
| M23109.1 | - | 5 - 6 | 486 | 105 | ||
More Information
Set Membership: |
|

F9
Genomic Map