Assay Details
| Gene Symbol: Rad51b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009014.3 | NP_033040.2 | 1 - 2 | 52 | 65 | |
| GenBank mRNA | AK032913.1 | - | 1 - 2 | 53 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032913.1, NM_009014.3 transcripts.
Set Membership: |
|

Rad51b
Genomic Map