Assay Details
| Gene Symbol: Rad51b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009014.3 | NP_033040.2 | 7 - 8 | 826 | 79 | |
| GenBank mRNA | AK032913.1 | - | 7 - 8 | 827 | 79 | |
| U92068.1 | - | 6 - 7 | 768 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad51b
Genomic Map