Assay Details
| Gene Symbol: Rad51b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252562.1 | NP_001239491.1 | 5 - 6 | 505 | 68 | |
| NM_009014.3 | NP_033040.2 | 5 - 6 | 523 | 68 | ||
| GenBank mRNA | AK015771.1 | - | 5 - 6 | 505 | 68 | |
| AK032913.1 | - | 5 - 6 | 524 | 68 | ||
| U92068.1 | - | 4 - 5 | 465 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad51b
Genomic Map