Assay Details
| Gene Symbol: Slc39a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026244.2 | NP_080520.2 | 6 - 7 | 1361 | 76 | |
| GenBank mRNA | AK080462.1 | - | 6 - 7 | 1343 | 76 | |
| AK090062.1 | - | 6 - 7 | 1354 | 76 | ||
| BC043112.1 | - | 6 - 7 | 1308 | 76 | ||
| BC072655.1 | - | 6 - 7 | 1337 | 76 | ||
| BC117754.1 | - | 6 - 7 | 955 | 40090588 | 76 | |
| BC117755.1 | - | 6 - 7 | 955 | 40090589 | 76 | |
| BC158056.1 | - | 6 - 7 | 1106 | 9088164 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976102 | BC158056.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976102 | BC117754.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976102 | BC117755.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652612 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920347 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943146 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290472 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202757 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329482 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 24467 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 199450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213660 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664181 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429089 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891086 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136759 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839937 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113360 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156292 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161924 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118023 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976102 | BC158056.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976102 | BC117754.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976102 | BC117755.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07681293 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161924 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118023 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07681360 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07681310 | - | - |
| Whole Mouse Genome G4122A | A_51_P482233 | CO040789.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482233 | BC043112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482233 | BC058649.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482233 | BC072655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482233 | NM_026244.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118023 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10160971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10106595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10171745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10209748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28576938 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10396904 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396898 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396901 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396899 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396902 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396897 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396903 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452765_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421250_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421249_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10396896 | - | - |
| Mouse Genome 430 2.0 Array | 1421250_at | - | - |
| Mouse Genome 430 2.0 Array | 1452765_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421249_at | - | - |
| Mouse Genome 430 2.0 Array | 1421250_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452765_at | - | - |
| Mouse Genome 430 2.0 Array | 1421249_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200005239.mm.1 | NM_026244.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005239.mm.1 | NM_026244.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005239.mm.1 | TC1200000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005237.mm.1 | NM_026244.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005237.mm.1 | NM_026244.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005237.mm.1 | TC1200000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005230.mm.1 | NM_026244.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005230.mm.1 | NM_026244.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005230.mm.1 | TC1200000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005231.mm.1 | NM_026244.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005231.mm.1 | NM_026244.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005231.mm.1 | TC1200000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005238.mm.1 | NM_026244.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005238.mm.1 | NM_026244.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005238.mm.1 | TC1200000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005235.mm.1 | NM_026244.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005235.mm.1 | NM_026244.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005235.mm.1 | TC1200000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005236.mm.1 | NM_026244.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005236.mm.1 | NM_026244.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005236.mm.1 | TC1200000741.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200005229.mm.1 | NM_026244.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005229.mm.1 | NM_026244.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200005229.mm.1 | TC1200000741.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452765_at | - | - |
| Mouse Expression Set 430 | 1421249_PM_at | - | - |
| Mouse Expression Set 430 | 1452765_PM_at | - | - |
| Mouse Expression Set 430 | 1421250_at | - | - |
| Mouse Expression Set 430 | 1421249_at | - | - |
| Mouse Expression Set 430 | 1421250_PM_at | - | - |
| Murine Genome U74v2 Set | 102232_PM_at | - | - |
| Murine Genome U74v2 Set | 102232_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452765_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421250_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452765_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421250_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421249_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421249_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421250_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421249_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421249_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421250_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452765_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452765_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102450133 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3940348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106200048 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2544111 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2460491 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106200048 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106200048 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2544111 | - | - |

Slc39a9
Genomic Map