Assay Details
| Gene Symbol: Rps6kl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC024798.1 | - | 7 - 8 | 1368 | 5358742 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps6kl1| Gene Symbol: Rps6kl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC024798.1 | - | 7 - 8 | 1368 | 5358742 | 67 |
Set Membership: |
|