| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200006198.mm.1 |
NM_001199843.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006198.mm.1 |
NM_001199843.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006198.mm.1 |
NM_029601.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006198.mm.1 |
NM_029601.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006198.mm.1 |
TC1200000846.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200006199.mm.1 |
NM_001199843.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006199.mm.1 |
NM_001199843.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006199.mm.1 |
NM_029601.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006199.mm.1 |
NM_029601.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006199.mm.1 |
TC1200000846.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200006197.mm.1 |
NM_001199843.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006197.mm.1 |
NM_001199843.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200006197.mm.1 |
TC1200000846.mm.1 |
MTA TC |