Assay Details
| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199843.1 | NP_001186772.1 | 4 - 5 | 273 | 84 | |
| GenBank mRNA | BC034668.1 | - | 4 - 5 | 211 | 84 | |
| BC038915.1 | - | 4 - 5 | 262 | 5148723 | 84 | |
| CF751010.1 | - | 4 - 5 | 236 | 84 | ||
More Information
Set Membership: |
|

Ift43
Genomic Map