Assay Details
| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199843.1 | NP_001186772.1 | 5 - 6 | 313 | 64 | |
| NM_029601.3 | NP_083877.1 | 4 - 5 | 280 | 64 | ||
| GenBank mRNA | AK006115.1 | - | 4 - 5 | 278 | 64 | |
| BC034668.1 | - | 5 - 6 | 251 | 64 | ||
| BC038915.1 | - | 5 - 6 | 302 | 5148723 | 64 | |
| CF751010.1 | - | 5 - 6 | 276 | 64 | ||
More Information
Set Membership: |
|

Ift43
Genomic Map