Assay Details
| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199843.1 | NP_001186772.1 | 6 - 7 | 391 | 77 | |
| NM_029601.3 | NP_083877.1 | 5 - 6 | 358 | 77 | ||
| GenBank mRNA | AK006115.1 | - | 5 - 6 | 356 | 77 | |
| BC034668.1 | - | 6 - 7 | 329 | 77 | ||
| BC038915.1 | - | 6 - 7 | 380 | 5148723 | 77 | |
| BE987571.1 | - | 3 - 4 | 77 | |||
| CF751010.1 | - | 6 - 7 | 354 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ift43
Genomic Map