Assay Details
| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038915.1 | - | 1 - 2 | 68 | 5148723 | 70 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC038915.1 transcripts.
Set Membership: |
|

Ift43
Genomic Map