Assay Details
| Gene Symbol: Gstz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252555.1 | NP_001239484.1 | 1 - 2 | 339 | 66 | |
| GenBank mRNA | AB041613.1 | - | 1 - 2 | 339 | 66 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB041613.1,NM_001252555.1 transcripts.
Set Membership: |
|

Gstz1
Genomic Map