Assay Details
| Gene Symbol: Tmem63c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172583.2 | NP_766171.1 | 11 - 12 | 1023 | 73 | |
| GenBank mRNA | AK030062.1 | - | 11 - 12 | 1015 | 73 | |
| AK034410.1 | - | 10 - 11 | 946 | 73 | ||
| AK043536.1 | - | 10 - 11 | 944 | 73 | ||
| BC056936.1 | - | 11 - 12 | 1023 | 5695989 | 73 | |
| BC057088.1 | - | 11 - 12 | 1023 | 5695989 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem63c
Genomic Map