Assay Details
| Gene Symbol: Il24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053095.2 | NP_444325.2 | 3 - 4 | 420 | 62 | |
| GenBank mRNA | AF235006.1 | - | 2 - 3 | 486 | 62 | |
| AF333251.1 | - | 3 - 4 | 348 | 62 | ||
More Information
Set Membership: |
|

Il24
Genomic Map