Assay Details
| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159301.1 | NP_001152773.1 | 4 - 5 | 551 | 101 | |
| GenBank mRNA | AK146044.1 | - | 4 - 5 | 507 | 101 | |
| U55060.1 | - | 4 - 5 | 477 | 101 | ||
More Information
Set Membership: |
|

Lgals9
Genomic Map