Assay Details
| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159301.1 | NP_001152773.1 | 5 - 6 | 586 | 78 | |
| GenBank mRNA | AK146044.1 | - | 5 - 6 | 542 | 78 | |
| U55060.1 | - | 5 - 6 | 512 | 78 | ||
More Information
Set Membership: |
|

Lgals9
Genomic Map