Assay Details
| Gene Symbol: Tnfaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159392.1 | NP_001152864.1 | 1 - 2 | 177 | 69 | |
| GenBank mRNA | AK147809.1 | - | 1 - 2 | 85 | 69 | |
| AK168386.1 | - | 1 - 2 | 69 | 69 | ||
| AK168805.1 | - | 1 - 2 | 85 | 69 | ||
| AK172287.1 | - | 1 - 2 | 93 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK147809.1, AK168386.1, AK172287.1, NM_001159392.1 transcripts.
Set Membership: |
|

Tnfaip1
Genomic Map